Learn More
Invitrogen™ PRIMER 3' AMINE
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
Specifications
Specifications
| Description | 3' Amine Primer, 200 nmol |
| Product Type | 3' Amine Primer |
| Primer | 3' Labeled |
| Quantity | 200 nmol |