missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PRIMER 3' BIO 200 NMOL

Product Code. 10174983
Change view
Click to view available options
Quantity:
200 nmol
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10174983 200 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10174983 Supplier Invitrogen™ Supplier No. 10626075
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Custom Primer

Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Normal or Reversed-phase Chromatography, HPLC or PAGE
Description Custom 3' BIO Primer, 200 nmole, Purified by Normal or Reversed-phase Chromatography, HPLC or PAGE, Used to Design a Custom DNA Oligonucleotide, With any of a Variety of 3' and 5' Modification
Product Type Custom 3' BIO Primer
Quantity 200 nmol
Grade HPLC
For Use With (Application) Design a Custom DNA Oligonucleotide, with any of a variety of 3' and 5' Modification
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.