Learn More
Invitrogen™ PRIMER, 5'ALEXA 514 200 NM
Product Code. 10355699
Click to view available options
:
200 nmol
Product Code. 10355699
Brand: Invitrogen™ 100007344
Custom Primer
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
TRUSTED_SUSTAINABILITY
Specifications
Description 5' Alexa 514 Primer, 200 nmol
Product Type 5' Alexa 514 Primer
Primer 5' Labeled
Quantity 200 nmol