Learn More
Invitrogen™ PRIMER, 5'ALEXA 660 50 NMOL
Click to view available options
:
50 nmol
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
Specifications
Specifications
| Description | 5' Alexa 660 Primer, 50 nmol |
| Product Type | 5' Alexa 660 Primer |
| Primer | 5' Labeled |
| Quantity | 50 nmol |