Learn More
Invitrogen™ PRIMER 5'BIOTIN 25 NMOL
Product Code. 10645733
Change view
Click to view available options
:
25 nmol
Product Code.
10645733 25 nmol
1 options
Product Code. 10645733 Supplier Invitrogen™ Supplier No. 10626133
Custom Primer
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
TRUSTED_SUSTAINABILITY
Specifications
Description 5' Biotin Primer, 25 nmol
Product Type 5' Biotin Primer
Content And Storage Room Temperature
Primer 5' Labeled
Quantity 25 nmol