Learn More
Invitrogen™ PRIMER 5' BIOTIN 50 NMOL
Custom Primer
Brand:  Invitrogen™ 10626018
76.50 EUR valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Additional Details : Weight : 0.00100kg
Product Code. 10434752
  • 102.00 € / Each
Estimated Shipment: 07-06-2024
to see stock.
Description
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
Specifications
Specifications
5' Biotin Primer, 50 nmol
5' Biotin Primer
5' Labeled
50 nmol