Learn More
Invitrogen™ PRIMER 5' FLO 200 NMOL
Custom Primer
Brand: Invitrogen™ 10918035
69.75 EUR valid until 2024-06-28
Use promo code "21649" to get your promotional price.
Additional Details : Weight : 0.45000kg
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).Specifications
5' FLO Primer, 200 nmol | |
5' FLO Primer | |
5' Labeled | |
200 nmol |