Learn More
Invitrogen™ PRIMER 5' HEX 200 NMOL
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
Specifications
Specifications
| Description | 5' HEX Primer, 200 nmol |
| Product Type | 5' HEX Primer |
| Primer | 5' Labeled |
| Quantity | 200 nmol |