Learn More
Emma Safety Footwear Primus Safety Shoes
Product Code. 15999091
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Size:
37
48
49
missing translation for 'unitSize':
1 pair
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size missing translation for 'unitSize'
15999091 48 1 pair
15989091 37 1 pair
15909101 49 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Product Code. 15999091 missing translation for 'mfr' Emma Safety Footwear missing translation for 'supplierNo' MM93986848
  • ONLINE EXCLUSIVE PRICE
    196.35 €
    122.85 € / 1 pairs
    Save 73.50 €
    37% Off
Estimated Shipment: 24-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Surprisingly light yet provides a strong and supportive fit. Waterproof even when fully submerged, thanks to its state-of-the-art Sympatex™ uppers. The all-weather safety shoe offers outstanding foot protection in all seasons.
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather / Waterproof construction
  • Waterproof Sympatex™ uppers
  • GripForce™ Duo Heat Resistant Outsole
  • Sanitized
Specifications
Certifications/Compliance EN ISO 20345:2011
Closure Type Laces
Color Black, Gray
Design Primus
Liner Material Hydro-Tec™ Sanitized Silver / Sympatex™
Material Leather
Size 48
Toe Material Steel