missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ NanoDrop™ Lite Plus Printer Accessory

Product Code. 31921093 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
NanoDrop Lite Plus Spectrophotometer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
31921093 NanoDrop Lite Plus Spectrophotometer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31921093 Supplier Thermo Scientific™ Supplier No. NDLPPRNTRMOD
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Use an optional printer with your NanoDrop™ Lite Plus Spectrophotometer. Print data on freezer-compatible labels.

Use an optional printer with your NanoDrop™ Lite Plus Spectrophotometer. Print data on freezer-compatible labels.

Print sample data on a cryogenic label

  • Sample number
  • Date
  • Time of Measurement
  • Sample Name
  • Assay
  • Concentration
  • Purity Ratio

For use with the NanoDrop Lite Spectrophotometer

Specifications

Description Printer Accessory for NanoDrop Lite Plus
For Use With (Equipment) NanoDrop Lite Plus Spectrophotometer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.