Learn More
Thermo Scientific™ Printers for CryoMed™ and CryoPlus™ Freezers
Product Code. 11746449 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Thermal Data Printer
Thermal Printer Paper
Voltage:
220 V
Not Applicable
Unit Size:
Each
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Voltage unitSize
11746449 Thermal Printer Paper Not Applicable Pack of 5
11721579 Thermal Data Printer 220 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11746449 Supplier Thermo Scientific™ Supplier No. 4000566
  • ONLINE EXCLUSIVE PRICE
    228.00 €
    213.00 € / Pack of 5
    Save 15.00 €
    6% Off
 Restricted Item
In Stock
Add to basket
Add to basket
Enhance the functionality for Thermo Scientific™ CryoMed™ controlled-rate freezers with this optional printer.
Enhance the functionality for Thermo Scientific™ CryoMed™ controlled-rate freezers with Thermo Scientific™ Thermal Printers for CryoMed and CryoPlus™ Freezers. These printers are available in 120V or 220V models, and include cable assembly and RS-232 connector kit.
  • Model 4000565 - 120V, 60Hz printer with cable assembly and RS-232 connector kit
  • Model 4000665 - 220V, 50Hz printer with cable assembly and RS-232 connector kit
  • Model 4000566 - Thermal printer paper (5 rolls/package)
TRUSTED_SUSTAINABILITY
Specifications
Type Thermal Printer Paper
For Use With (Equipment) All CryoMed Controlled-Rate Freezers
Voltage Not Applicable