Learn More
Invitrogen™ Human PRKACB (PRKAC beta), GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Recombinant full-length human PRKACB was expressed by baculovirus in Sf9 insect cells using a N-terminal GST tag. Validated in LanthaScreen™ Kinase Binding Assay Protocols available for simple use and optimized performance Multiple pack sizes available for convenient use Related Product:LanthaScreen™ Eu Kinase Binding Assay Protein Form: Recombinant, Full Length Assay Entry: Biochemical binding/kinetic The gene accession number for PRKACB (PRKAC beta) is NP_002722.1.
Specifications
Specifications
| Accession Number | P22694 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50mM tris HCl with 150mM NaCl, 25% glycerol, 0.1mM EGTA, 0.1mM PMSF, 0.25mM DTT, 0.1mM EDTA and no preservative; pH 7.5 |
| Gene ID (Entrez) | 5567 |
| Molecular Weight (g/mol) | 66.9 kDa |
| Name | Human PRKACB (PRKAC beta), GST Tag |
| pH Range | 7.5 |
| Purification Method | Purified |
| Quantity | 10 μg |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.