Learn More
Invitrogen™ Human PRKCB2 (PKC beta II), GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length, Active The gene accession number for PRKCB2 (PKC beta II) is NP_002729.2.
Specifications
Specifications
| Accession Number | P05771 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50mM tris HCl with 15mM glutathione, 100mM NaCl, 20% glycerol, 5mM DTT and no preservative; pH 8 |
| Gene ID (Entrez) | 5579 |
| Molecular Weight (g/mol) | 103.7 kDa |
| Name | Human PRKCB2 (PKC beta II), GST Tag |
| pH Range | 8 |
| Purification Method | Purified |
| Quantity | 10 μg |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.