Learn More
Chemsplash Pro+4 (Sterile Irradiated) 67gsm C/All Taped Seams
GREENER_CHOICE
Product Code. 30058241
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 25
Product Code. 30058241
Brand: Chemsplash 2767.400.004
Only null left
Add to basket
Add to basket
Gamma Irradiated Chemsplash Pro +4 Disposable Coverall is made from a 67 gsm microporous laminated Cat III Type 4B, 5B and 6B fabric.
Gamma Irradiated Chemsplash Pro +4 Disposable Coverall
  • 67GSM Microporous Laminate Fabric
  • Sterile Irradiated, Double Bagged and Detex Sticker
  • Fully Taped Seams
  • Three Piece Hood
  • Elasticated Cuffs with Thumb Loops
  • Elasticated Hood, Back and Ankles
  • Two Way Zip with Zip Flap
  • Silicone and Latex Free
  • Low Linting, Anti-Static
  • Conforms to ISO 14644-1:2015 Cleanroom 5 - 9
Description: The Gamma Irradiated Chemsplash Pro +4 Disposable Coverall is made from 67GSM microporous laminated Cat III Type 4B, 5B and 6B fabric. All seams are fully taped, offering a stronger and more effective barrier against liquids and particulate matter. This coverall is anti-static to EN1149-5:2008 and resistant to infectious agents under EN14126, ensuring compliance with viral and bacterial infection protection. It is ideal for use in pharmaceutical, cleanroom, semiconductor manufacturing, and infection contamination control environments. This coverall conforms to ISO 14644-1:2015 Cleanroom Class 5-9, offering maximum protection for highly sensitive environments.
Specifications
Product Type Cleanroom Coveralls
Size Small
Gender Unisex
Material Microporous Fabric
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Knit, Thumb Loop
Neck Style Hooded
Hood Type 3-panel
Closure Type Ties
Seam Type Stitched, Taped
Sterility Sterile
Resistance Liquid
Latex Free Yes
No. of Pockets None
Cleanroom Class Class 100-100,000 (ISO 5-8)