Learn More
Chemsplash Jet Spray 88 Type 3/4 5/6 Coverall

Description
Chemsplash Jet Spray 88 Type 3B/4B 5B/6B Coverall
- 88GSM Heavyweight Microporous Non-Woven Fabric
- Ultrasonically Tape Welded Seams
- Self Adhesive Chin Strap for Optimum Protection
- Elasticated Hood, Half Waist, Cuffs, and Ankles
- Convenient Thumb Loops at Wrist
- Single-Way Zip with Bi-Folding Self Adhesive Flap
- Silicone and Latex Free
- Non Linting Fabric
- Anti-Static
- PFAS Free
Description: The Chemsplash Jet Spray 88 Type 3B/4B 5B/6B Coverall is designed to provide high resistance against chemical permeation, offering exceptional protection against even the most corrosive chemicals. Made from a durable 88gsm heavy-weight Polypropylene/Polyethylene multilayer laminated material, it also provides protection against biological hazards, including viruses, bacteria, and blood-borne pathogens, according to EN14126. In addition, the coverall offers protection against particulate radioactive contamination (level 2) as per EN1073-2 standards. The design includes a self-adhesive chin strap, elasticated hood, waist, cuffs, and ankles, and thumb loops for increased comfort and protection. This coverall is perfect for environments where high levels of chemical and biological protection are required.
Specifications
Specifications
| Product Type | Cleanroom Coveralls |
| Size | 2X-Large |
| Gender | Unisex |
| Material | Microporous Composite Fabric |
| Color | Yellow |
| Disposable | Single-use |
| Waist Style | Elastic |
| Ankle Style | Elastic |
| Cuff Style | Elastic, Thumb Loop |
| Neck Style | Hooded |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.