Learn More
Chemsplash Pro 63 Coverall (Sterile Irradiated)
GREENER_CHOICE
Product Code. 30058253
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 25
Product Code. 30058253
Brand: Chemsplash 2783.400.004
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Pro 63 Coverall Sterile Irradiated is made from a strong yet breathable 63 gsm microporous laminated fabric.
Chemsplash Pro 63 Sterile Irradiated Coverall
  • 63GSM Thermal Weld Microporous Laminate Fabric
  • Sterile Irradiated, Double Bagged and Detex Sticker
  • Conforms to ENISO14644-1 Class 5
  • Elasticated 3-Piece Hood, Knitted Cuffs
  • Two Way Zip, Adhesive Zip Flap
  • Internal Hang Loop
  • Non Linting
  • Silicone and Latex Free
  • Anti-Static
  • Minimum Quantity 700 Pcs Applies
Description: The Chemsplash Pro 63 Coverall is made from strong yet breathable 63gsm microporous laminated fabric. It is a high-quality and robust Cat III Type 5 and 6 suit that meets stringent industry requirements. The Chemsplash Pro fabric is non-linting and anti-static to EN 1149-5:2008, making it ideal for applications where contamination from garment fibers must be prevented. Featuring a knitted cuff and two-way zip for comfort and fit, the zip is sealed with an adhesive cover flap for added protection. The coverall is gamma-irradiated for sterilization, double-bagged, and clearly marked, ensuring readiness for use right out of the bag. This is a premium Cat III Type 5 and 6 coverall designed for single use in cleanroom environments, offering low linting and shedding.
Specifications
Product Type Cleanroom Coveralls
Size Small
Gender Unisex
Material Microporous Fabric
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Knit
Neck Style Hooded
Hood Type 3-panel
Closure Type Ties
Seam Type Stitched
Sterility Sterile
Resistance Liquid
Latex Free Yes
No. of Pockets None
Cleanroom Class Class 100-100,000 (ISO 5-8)