missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pro-Detect™ Rapid Antibody Isotyping Assay Kit, mouse

Product Code. 15900533 Shop All Thermo Scientific Products
Click to view available options
Quantity:
10 tests
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 15900533

Brand: Thermo Scientific™ A38550

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

10 assays

Pro-Detect™ Rapid Antibody Isotyping Assay Kit is a single-use, dipstick lateral flow kit for quick, easy determination of monoclonal antibody class and subclass identity

  • Uses membrane-based strips coated with dye-bound conjugated antibodies that provide a visual, color readout of the monoclonal antibody isotype within ten minutes.
  • Lateral flow strips can be used early in and throughout expression to monitor and confirm expression of desired protein-tagged target
  • Simple, fast method—dilute sample using provided diluent and dip in the lateral flow strip for visual results within 5 to 10 minutes
  • Sensitive—detect protein tags in supernatant or cell lysate samples down to low μg/mL concentration
  • Specific—assay does not cross-react with common lysis and protein preparation reagents
  • Reliable—results are similar to other antibody immunoassays, including western blot and ELISA
  • Long shelf life—stable for at least one year at 4°C

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 10 Rapid Antibody Isotyping Assay Strips 1
• 10 Rapid Antibody Isotyping Assay Strips 2
• Sample Diluent, 4 mL

Sufficient for 10 isotype determinations

Store at 2–8°C.
Detectable Analytes IgG1, IgG2a, IgG2b, IgG3, IgA, IgM; Kappa and lambda light chains
Assay Isotyping Assay
Detection Method Chromogenic
Product Line Pro-Detect
Product Type Assay
Sample Type Ascitic Fluid, Cell Culture Media
Specificity Antibody
Sufficient For 10 tests
Target Species Mouse
For Use With (Application) Lateral Flow Rapid Assay
Quantity 10 tests
Shipping Condition Wet Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.