missing translation for 'onlineSavingsMsg'
Learn More

Pro-Lab Diagnostics™ Microbank™ Bacterial and Fungal Preservation System

Product Code. 17759704
Change view
Click to view available options
Closure Color:
Green
Unit Size:
Pack of 80
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Color Unit Size
17759704 Green Pack of 80
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17759704 Supplier Pro-Lab Diagnostics™ Supplier No. PL.170C/G
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

Designed to simplify the storage and retrieval of bacterial and fungal cultures. Prolab Diagnostics™ Microbank™ Bacterial and Fungal Preservation System is a ready-to-use system.

  • Incorporates treated beads and a special cryopreservative solution
  • Each vial contains 25 beads, providing repeated identical cultures from the original organism
  • Specially formulated preservative ensures longer survival of fastidious cultures and higher quantitative recoveries
  • 80 Vials supplied in a freezer box with“see through”cover
  • Available in 5 cap/bead colors or mixed (16 vials each of blue, green, light blue, red, and yellow)

Order Info

System supplied as box of 80 vials

Specifications

Product Type Cryogenic Storage Vial
Closure Color Green
For Use With (Application) Designed to simplify the storage and retrieval of bacterial and fungal cultures
Quantity 80 Pk.
Storage Requirements -70°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.