missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Pro-Q™ Emerald 300 Lipopolysaccharide Gel Stain Kit

Product Code. 10033292
Click to view available options
Quantity:
1 kit
Unit Size:
9g
This item is not returnable. View return policy

Product Code. 10033292

Brand: Invitrogen™ P20495

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pro-Q™ Emerald 300 Lipopolysaccharide Gel Stain Kit

Molecular Probes' proprietary Pro-Q Emerald 300 and Pro-Q Emerald 488 Glycoprotein Stain Kits provide the most advanced technology available for detection of glycoproteins in gels and on blots. Gel staining is rapid and very sensitive. In less than three hours, it is possible to detect as little as 300 pg of glycoprotein per band, depending on the degree of glycosylation, making these stains at least 50-fold more sensitive than standard fuchsin staining.
TRUSTED_SUSTAINABILITY

Specifications

Detection Location In-Blot Detection, In-Gel Detection
Detection Method Fluorescence
Label or Dye Pro-Q Emerald 300
Product Type Lipopolysaccharide Stain Kit
Product Line PRO-Q
Quantity 1 kit
Target Molecule Lipopolysaccharide
compliance-icons compliance-icons
Product Identifier
  • Pro-Q Emerald 300 Lipopolysaccharide Gel Stain Kit
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Periodic acid

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.