missing translation for 'onlineSavingsMsg'
Learn More

Techsit™ Pro-Tech Laboratory Low Chair

Product Code. 16543904
Encompass_Preferred
Change view
Click to view available options
Color:
Black
Gray
With Armrest:
No
Yes
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color With Armrest unitSize
16543904 Black No Each
16553904 Black Yes Each
16563904 Gray No Each
16573904 Gray Yes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16543904 Supplier Techsit™ Supplier No. TS7001B
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Advanced laboratory low chair is fully adjustable with an adjustable foot ring.

  • Vinyl low chair on casters
  • Antimicrobial vinyl that is easy to clean
  • Fully adjustable seat height, back height, and seat depth
  • Ergonomically designed chair that automatically adjusts to the user
  • Cleanroom Class 7 suitable

Specifications

Product Type Laboratory Seat
Color Black
Adjustable Yes
Autoclavable Not Autoclavable
Back Style Ergonomic
Finish Vinyl
Frame Material Steel
Lift Type Gas Lift
Load Bearing Capacity (Metric) 150 kg
Seating Height Desk Height
Upholstery Vinyl
Warranty 5 Year
Weight (Metric) 16 kg
Wheel Type Casters
Width (Metric) 490 mm
With Armrest No
With Backrest Yes
With Casters Yes
Footrest No
With Lumbar Support Yes
With Tilt Yes
Material Vinyl
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.