missing translation for 'onlineSavingsMsg'
Learn More

Proadifen hydrochloride, 98%, BLD Pharm™

Product Code. 31097441
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31097441 250 mg
31096983 1 g
31097202 100 mg
31096376 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31097441 Supplier BLD Pharm Supplier No. BD149084250mg
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 62-68-0
Molecular Formula C23H32ClNO2
MDL Number MFCD00055151
Quantity 250 mg
SMILES CCCC(C1=CC=CC=C1)(C2=CC=CC=C2)C(OCCN(CC)CC)=O.[H]Cl
Molecular Weight (g/mol) 389.96
Percent Purity 98%
Chemical Name or Material Proadifen hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.