missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Probe Set Diluent QT

Product Code. 15922334
Change view
Click to view available options
Quantity:
12 mL
Unit Size:
12mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15922334 12 mL 12mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15922334 Supplier Invitrogen™ Supplier No. QVT0510
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Probe Set Diluent QT, 12 mL is an aqueous solution containing formamide, detergent, and blocker.

Probe Set Diluent QT, 12 mL is an aqueous solution containing formamide, detergent, and blocker. It is an individual component of the ViewRNA™ ISH Tissue Assay Kits, 2-Plex, 24 assays.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2°C to 8°C
Format Kit
Product Type Probe Set Diluent
Quantity 12 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.