missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Probenecid

Product Code. 18752614
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18752614 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18752614 Supplier R&D Systems™ Supplier No. 4107/50
  • ONLINE EXCLUSIVE PRICE
    92.08 €
    82.95 € / 50mg
    Save 9.13 €
    9% Off
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. MRP inhibitor

Probenecid is an inhibitor of multidrug resistance-associated proteins (MRP). Probenicid inhibits OAT3, organic acid transport in the kidney and other organs and exhibits inhibitory activity against pannexin 1 channels (IC50 ∼ 150 μM). Probenecid potently inhibits SARS-CoV-2 replication in NHBE and Vero E6 cells (IC50 = 1.3 nM and 750 nM, respectively) and reduces lung virus titers in vivo. Probenicid also inhibits influenza A virus replication in vitro and in vivo.
TRUSTED_SUSTAINABILITY

Specifications

CAS 57-66-9
Quantity 50 mg
Product Line Tocris
Molecular Formula C13H19NO4S
Formula Weight 285.36
Purity >98%
Molecular Weight (g/mol) 285.36
Chemical Name or Material Probenecid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.