missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ProBond™ Nickel-Chelating Resin
Descrizione
Includes
The ProBond resin is provided as a 50% slurry in 20% ethanol.
- ProBond™ Nickel-Chelating Resin is a nickel-charged affinity resin used to purify recombinant proteins containing a polyhistidine (6xHis) sequence
- Proteins bound to the resin may be eluted with either low pH buffer or by competition with imidazole or histidine
- One-step purification can be performed under both native and denaturing conditions
- ProBond™ Resin uses the chelating ligand iminodiacetic acid (IDA) coupled to a highly cross-linked 6% agarose resin
- Suitable for use in FPLC, batch, and gravity flow applications
- Guaranteed stable for 6 months when properly stored at 4°C
Specifica
Specifica
| Content And Storage | The ProBond™ resin is pre-charged and able to bind 1-5 mg of recombinant protein per 1 ml of resin. It is provided as a 50% slurry in 20% ethanol. The resin will appear blue in color when charged with Ni2+ . Store at +4°C. ProBond™ resin is guaranteed stable for 6 months when properly stored. |
| Stationary Phase | Nickel-Chelating |
| Product Line | ProBond |
| Type | Resin |
| Quantity | 50 mL |
| Form | Suspension |
For Research Use Only. Not for use in diagnostic procedures.
Titolo del prodotto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?