missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Procarbazine hydrochloride

Product Code. 15537729 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15537729 250 mg 250mg
15527729 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15537729 Supplier Thermo Scientific Chemicals Supplier No. 461182500
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 366-70-1 | C12H19N3O·HCl | 257.76 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 366-70-1
Molecular Formula C12H19N3O·HCl
Molecular Weight (g/mol) 257.76
InChI Key DERJYEZSLHIUKF-UHFFFAOYSA-N
Synonym procarbazine hydrochloride, procarbazine hcl, matulane, nathulane, natunalar, mih hydrochloride, pcb hydrochloride, procarbazine.hcl, ibenzmethyzine hydrochloride, natulan hydrochloride
PubChem CID 9703
ChEBI CHEBI:71428
IUPAC Name 4-[(2-methylhydrazinyl)methyl]-N-propan-2-ylbenzamide;hydrochloride
SMILES CC(C)NC(=O)C1=CC=C(C=C1)CNNC.Cl

Specifications

Loss on Drying 2% max.
Infrared Spectrum Authentic
Quantity 250 mg
Formula Weight 257.76
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Procarbazine hydrochloride
compliance-icons compliance-icons
Product Identifier
  • Procarbazine hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1A
Hazard Statement
  • H360-May damage fertility or the unborn child.
  • H350-May cause cancer.
  • H341-Suspected of causing genetic defects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.