missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ProcartaPlex™ Human VIP Competitive Simplex Kit

Product Code. 31942166
Change view
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31942166 96 Tests 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31942166 Supplier Invitrogen™ Supplier No. EPX01012371901
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The ProcartaPlex Human VIP Competitive Simplex Kit measures VIP protein and is based on competitive ELISA technique, where sample Testosterone competes with a labeled tracer for antibody binding, resulting in an inverse signal proportional to cortisol concentration.

The ProcartaPlex Human VIP Competitive Simplex Kit measures VIP protein and is based on competitive ELISA technique, where sample Testosterone competes with a labeled tracer for antibody binding, resulting in an inverse signal proportional to cortisol concentration.

This kit can be used for single-target quantification or combined with other simplex assays to create a panel. ProcartaPlex Competitive Simplex kits are compatible with each other and can also be combined with sandwich assays. Please use the Panel Configurator to verify the compatibility of your desired panel.

A ProcartaPlex Basic Kit (sold separately) is required to run the assay, as Simplex kits include only assay-specific components and do not contain any buffer or plate.

About ProcartaPlex Assays for the Luminex Platform

ProcartaPlex assays are bead-based immunoassays designed for multiplex protein analysis on Luminex™ xMAP instruments. They enable the simultaneous detection and quantification of multiple protein targets from a single, small-volume sample, helping researchers save time, sample, and cost compared to traditional single-analyte ELISAs.

Depending on the analyte, ProcartaPlex assays are based on either sandwich ELISA or competitive immunoassay principles.

  • Sandwich assays use two highly specific antibodies binding to different epitopes of the target protein.
  • Competitive assays measure small molecules or analytes with limited epitopes, where the sample analyte competes with a labeled tracer for antibody binding, resulting in an inverse signal.

How the technology works

ProcartaPlex assays use Luminex xMAP technology, in which microscopic magnetic beads are internally dyed with precise ratios of red and infrared fluorophores. Each bead population has a unique spectral signature and is coated with antibodies specific to a single target. This allows different analytes to be measured in parallel within the same well.

Unlike traditional ELISA, where antibodies are immobilized on a plate, ProcartaPlex reagents are free-floating in solution, improving reaction kinetics and enabling efficient multiplexing with the following benefits:

  • Multiplexing efficiency—measure up to 80 protein targets simultaneously
  • Low sample volume—as little as 25 μL for serum or plasma, or 50 μL for cell culture supernatant
  • Time-saving workflow—results in approximately four hours
  • High Throughput - 96-well and even 384-well assay formulations are available helping conserve valuable samples while increasing assay efficiency.

ProcartaPlex multiplex panels and simplex assays are available for six (species: human, mouse, rat, nonhuman primate, porcine, and canine).

For more information and a full list of available targets, visit thermofisher.com/procartaplex.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Magnetic Bead-based Assay
Quantity 96 Tests
Species Human
Combinability Combinable
Bead Type Magnetic
Bead Region VIP [15]
Assay Sensitivity As determined for Lot 1: 5.675 pg/mL
Assay Range As determined for Lot 1: 25,400 to 6.2 pg/mL
Format Simplex Kit
For Use With (Equipment) Luminex Instruments
Gene VIP
Gene Alias PHM27
Gene ID (Entrez) 7432
Interassay CV As determined for Lot 1: 7.4 pg/mL
Intraassay CV As determined for Lot 1: 7.1 pg/mL
Product Line ProcartaPlex
Sample Type Serum, Plasma, Cell Culture Supernatants, Cerebrospinal Fluid
Sample Volume Serum, Plasma, CSF: 25 μL, CCS: 50 μL
UniProt ID P01282
Shipping Condition Wet Ice
Research Area Endocrinology
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.