missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ProcartaPlex™ Platinum Assay Buffer for Plasma Samples
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
Description
ProcartaPlex Platinum Assay Buffer for Plasma Samples is used for dilution of plasma samples and gives spike- and dilution-recovery results in the range of 70–130%.
ProcartaPlex Platinum Assay Buffer for Plasma Samples is included in our Platinum ProcartaPlex Human Basic Kit for Plasma Samples (Cat.No. EPXP010-10421-901) and off-the-shelf Platinum panels. It is offered here as a stand-alone item.
Specifications
Specifications
| Product Type | Buffer |
| Quantity | 10 mL |
| Species | Human |
| For Use With (Equipment) | Luminex™ Instruments |
| Product Line | ProcartaPlex |
| Sample Type | Plasma |
| Content And Storage | Store in refrigerator 2°C to 8°C |
| Shipping Condition | Wet Ice |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction