missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ProcartaPlex™ Platinum Assay Buffer for Serum Samples

Product Code. 16336672
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16336672 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16336672 Supplier Invitrogen™ Supplier No. EPXP11113000
  • ONLINE EXCLUSIVE PRICE
    188.00 €
    180.48 € / 10mL
    Save 7.52 €
    4% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ProcartaPlex Platinum Assay Buffer for Serum Samples (1X) is used for dilution of serum samples and gives spike- and dilution-recovery results in the range of 70 to 130%.

ProcartaPlex Platinum Assay Buffer for Serum Samples (1X) is used for dilution of serum samples and gives spike- and dilution-recovery results in the range of 70–130%.

ProcartaPlex Platinum Assay Buffer for Serum Samples (1X) is offered here as a stand-alone item for Platinum ProcartaPlex panels.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Buffer
Quantity 10 mL
Species Human
Form Liquid
Product Line ProcartaPlex
Content And Storage Store in refrigerator 2°C to 8°C
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.