missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Procion Blue HB

Product Code. 16498768
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498768 10 mg 10mg
16458916 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16498768 Supplier MedChemExpress Supplier No. HYD096510mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Procion Blue HB (Reactive Blue 2) is a purinergic antagonist.

Specifications

Content And Storage 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Quantity 10 mg
Shipping Condition Room temperature
Solubility DMSO : ≥ 110 mg/mL (142.09 mM) ∣ H2O : 10 mg/mL (12.92 mM; Need ultrasonic) ∣ Ethanol : 1 mg/mL (1.29 mM; Need ultrasonic)
Target HBV
Product Type Dye Reagents
Molecular Weight (g/mol) 774.16 g/mol
CAS 12236-82-7
Synonym Reactive Blue 2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.