missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva EPS 3501 High Voltage Power Supplies for Electrophoresis Units
Cover the wide range of electrophoresis and blotting applications. Designed for performance, EPS Power Supplies can be relied on for security and reproducibility
Brand: Cytiva 18-1130-05
Description
- Feature membrane keypads and clear digital displays for ease of programming
- Timed or continuous runs from 1 min. to 500 hrs., with end-of-run alarm
- Automatic parameter limit crossover prevents overheating, protects experiments and equipment
- Two pairs of power outlets permit duplicate parallel runs
- Automatic recovery after power failure
Four available modes (depending on model)
- Constant current mode is recommended for blotting to prevent overheating
- Constant voltage mode is recommended for other applications, such as nucleic acid submarine and mini-vertical gel electrophoresis
- Constant power mode is designed for constant temperature runs without a circulating cooler
- Linear voltage ramp mode brings proteins to focused equilibrium in IPG strips quickly without danger of overheating
EPS 3501
- Primarily designed for manual DNA sequencing and IEF applications
- Also suitable for many electrophoresis and blotting applications
- Constant voltage, constant current, or constant power modes
- Stores and recalls three protocols
- Primarily designed for 2-D electrophoresis and IEF using ImmobilineTM, but also suitable for many other electrophoresis applications
- For high-voltage applications requiring advanced programming
- Constant voltage, constant current, constant power, and linear voltage ramp modes
- Stores and recalls 9 protocols, each with 9 phases and phase-end alarms
- Programmed automatic shut-off or phase end based on time, volt-hour/milliampere-hour
Informations sur la commande
D x W x H: 12 1/4 x 9 3/4 x 3 3/4 in. (31.0 x 25 x 9.5cm)
Spécification
EN61010-1, CE 89/336/EEC (EMC directive), CE 73/23/EEC (LV directive) | |
1 min. to 500 hr. | |
Digital | |
106 kPa | |
39.2°F to 104°F | |
12.25 x 9.75 x 3.75 in. (31 x 25 x 9.5 cm) | |
31 cm | |
25 cm | |
9.5 cm | |
3.8 kg | |
3500 V | |
400 mA |
Immobiline Dry strip, IPG, Electrophoresis, Basic Blotting | |
EPS 3501 XL Power Supply | |
0 to 0.95% | |
0.2 cm | |
4°C to 40°C | |
12.25 in. | |
9.75 in. | |
3.75 in. | |
8.3 lb. | |
102 to 120 V / 200 to 240 V; 50/60 Hz | |
200 w | |
50/60 Hz |