Learn More
Sartorius Arium™ pro Ultrapure Water Systems
Intuitive menu navigation ensures error-free operation in consistent water quantity.
Brand:  Sartorius H2OPRO-VF-B
Code : NEW
Additional Details : Weight : 0.20000kg
Product Code. 11725016
  • 9790.00 € / Each
Estimated Shipment: 02-09-2024
to see stock.
Add to basket
Description
Description
  • pro VF Wall-Mounted System
  • High Efficiency UV Lamp (185/254 nm) & TOC Monitoring
  • Eliminates bacteria and organics
  • Display with touch-function
  • Integrated real-time monitoring
  • Includes wall bracket for the system, UV Lamp and Ultrafilter
Specifications
Specifications
Ultrapure Water Systems
Arium pro Ultrapure Water System
wall bracket for the system, UV Lamp and Ultrafilter
Wall