missing translation for 'onlineSavingsMsg'
Learn More

uvex Profaprotect CN34 Chemical-protection Gloves

Product Code. 31910429 Shop All Uvex Products
Change view
Click to view available options
Size:
7
8
9
10
11
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
31910429 8 Each
31910419 7 Each
31910420 9 Each
31910416 10 Each
31910421 11 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31910429 Supplier uvex Supplier No. 6072008
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Flexible chemical protection glove with cotton flocking for protection against a wide range of chemicals.

  • The profaprotect CN34 offers excellent chemical resistance to a wide range of aggressive chemicals, cleaning agents, adhesives and solvents such as acetone, ketones, aldehydes, acids, bases and oils.
  • Its flexibility and sensitivity ensure a high level of wearer comfort.
  • Certified chemical protection from 11 of the 18 test chemicals.
TRUSTED_SUSTAINABILITY

Specifications

Applications Laboratory, Healthcare, Industrial
Color Orange
Latex Free Yes
Material NBR, Chloroprene
Product Type Chemical Protection Gloves
Size 8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.