Learn More
uvex Profastrong NF33 Chemical Protection Gloves
Product Code. 15396922 Shop All Uvex Products
Change view
Click to view available options
Size:
7
8
9
10
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15396922 9 Each
15386922 8 Each
15366922 10 Each
15376922 7 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15396922 Supplier uvex Supplier No. 6012203
Only null left
Add to basket
Edge
Add to basket
Sensitive NBR chemical protection gloves feature a flocked cotton lining for comfort and provide a high degree of mechanical abrasion resistance.
  • Sensitive NBR chemical protection gloves with flocked cotton lining for comfort
  • Embossed structure on the palm enhances grip capabilities in damp and wet areas
  • Nitrile rubber is resistant to oils, grease, acids, and alkalis
  • Gloves provide a tactile feel, secure fit, and mechanical abrasion resistance
  • Ideal for use in various industries including automotive, chemical, and printing
TRUSTED_SUSTAINABILITY
Specifications
Color Green
Latex Free Yes
Material NBR
Product Type Chemical-resistant Gloves
Size 9
EN388 Standard 4101X
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES AJKLOT
EN ISO 374 - 5 Bacteria, Fungi WITH Virus Protection
Glove Cuff Style Gauntlet
Liner Material Cotton
Series Profastrong NF33
Length (Metric) 33 cm