missing translation for 'onlineSavingsMsg'
Learn More

Profenofos-D3, TRC

Product Code. 30426849
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30426849 2.5 mg 2.5mg
30427123 10 mg -
30411964 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30426849 Supplier Toronto Research Chemicals Supplier No. TRCP7558842.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2140327-42-8
Molecular Formula C11 2H3 H12 Br Cl O3 P S
Molecular Weight (g/mol) 376.65
Synonym Profenofos D3
IUPAC Name 1-bromo-3-chloro-2,5,6-trideuterio-4-[ethoxy(propylsulfanyl)phosphoryl]oxybenzene
SMILES [2H]c1c([2H])c(OP(=O)(OCC)SCCC)c(Cl)c([2H])c1Br

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C11H15BrClO3PS/c1-3-7-18-17(14,15-4-2)16-11-6-5-9(12)8-10(11)13/h5-6,8H,3-4,7H2,1-2H3/i5D,6D,8D
Formula Weight 374.954
Chemical Name or Material Profenofos D3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.