Learn More
TFA Dostmann Professional Digital Thermometer with Penetration Probe P200
Product Code. 12647653
Change view
Click to view available options
Calibrated:
Yes
Unit Size:
Each
Product Code. Calibrated Unit Size
12647653 Yes Each
1 options
Product Code. 12647653 Supplier TFA Dostmann Supplier No. 30.1015
  • ONLINE EXCLUSIVE PRICE
    41.80 €
    30.56 € / Pack of 1
    Save 11.24 €
    26% Off
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Professional digital thermometer with penetration probe P200 is ideal for professional temperature control of liquid, paste-like and semi-solid materials. It displays the current, the highest and lowest reading temperature. It is jet-proof and HACCP compliant.
This functional thermometer is ideal for simultaneous measurement of indoor and outdoor temperatures. Thanks to the large display, the values are particularly easy to read. Controlling the indoor temperature is an important factor for a comfortable and healthy living environment and helps save energy. Even one degree less temperature can bring up to 6% reduction in heating costs. When placed at the window, the sensor cable is simply led outside to measure the outdoor temperature. The max.-min. memory with manual reset allows the observation of peak values over a self-selected period of time. The application possibilities of thermometers with sensor cables are manifold and not only limited to the outdoor temperature. Due to the large measuring range, the waterproof measuring sensor can also be used for temperature monitoring of the refrigerator and freezer or for measuring the water temperature in the aquarium.
  • Penetration probe (NCT, 110 mm) with approx. 60 cm cable
  • For temperature measurement of air, gas, liquids and paste-like substances
  • With integrated probe holder, pocket clip and stand
  • Large display
  • Jet-proof to IP65
  • Hold-, max.-min.-function, washable
  • Compliant with HACCP, ideal for professional use
  • Resolution 0.1 °C, precision ±1°C (-25...+150°C), otherwise ±1,5°C
Specifications
Material Plastic
Calibrated Yes
Display LCD