missing translation for 'onlineSavingsMsg'
Learn More

Ansell Profile 3000 Nitrile Examination Gloves

Product Code. 12930825
Change view
Click to view available options
Size:
2X-Small
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 100
Pack of 1000
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12930825 Medium Pack of 100
12910825 X-Small Pack of 100
12940825 Large Pack of 100
12970825 3X-Large Pack of 100
13178646 2X-Small Pack of 1000
12920825 Small Pack of 100
12950825 X-Large Pack of 100
12960825 2X-Large Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 12930825 Supplier Ansell Supplier No. PNA3103M
Stock Up Your Lab
Use promo code 28007
View all promotions
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Combine comfort and chemical resistance with Nitritex™ Profile 3000 nitrile exam gloves. Ambidextrous, latex-free glove offers chemical and puncture resistance.

  • Nitrile is resistant to a wide range of chemicals
  • Excellent puncture resistance
  • Beaded cuff provides easier donning and stability
  • Textured surface for increased grip
  • Powder-free and latex-free to eliminate Type I latex allergies
  • Wrist-length gloves measure 240mm/9.5 in.
  • Easy-access 100-glove dispenser box
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO 9001:2008, 13485:2003 and ISO 2859
Classification Class I Medical Device
Color White
Disposable Single-use
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Nitrile Examination Glove
Size Medium
Sterility Non-sterile
For Use With (Application) Premium examinations
Glove Cuff Style Beaded/Rolled
Inspected for Medical Use Yes
Model Profile 3000
Packaging Type Boxed
Quantity 100/Pk.
Series Profile 3000
Length (Metric) 24 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.