missing translation for 'onlineSavingsMsg'
Learn More

WTW™ ProfiLine™ Portable Conductivity Meters
GREENER_CHOICE

Product Code. 10414885
Change view
Click to view available options
Type:
Conductivity Meter Field Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
10414885 Conductivity Meter Field Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10414885 Supplier WTW™ Supplier No. 2CA101
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Obtain optimum measurements in natural waters and wastewater with this reliable conductivity portable meter.

  • Robust and waterproof meter for portable conductivity measurement
  • Reliable conductivity meter with automatic nIF temperature compensation according to DIN EN 27888 for routine measurements in natural waters and wastewater
  • Features automatic temperature compensation nlF
  • For professional field measurements requiring documentation and data transfer
TRUSTED_SUSTAINABILITY

Specifications

Includes Meter with carry case including 4-electrode conductivity cell TetraCon™ 325, standard solution 1413μS/cm at 25°C, stand, beaker, short instruction manual, CD-ROM and batteries
Accuracy (Conductivity) ±0.5% Full-scale ±1 LSD
For Use With (Application) For professional field measurements requiring documentation and data transfer
Cell Constant Fixed: 0.475cm–1/1cm–1, Calibratable: 0.450 to 0.500cm–1/0.585 to 715cm–1/0.800 to 1.200cm–1
Calibration Points 1, 2, 3-point WTW Technical and NIST Buffer
Range (Conductivity) 0 μS to 1000 mS
Resolution (Conductivity) 0.001, 0.01, 0.1 mS
Temperature Compensation Automatic
Range (Salinity) 0 to 70 ppt
Resolution (Salinity) 0.1 ppt
Accuracy (Salinity) ±0.2% Full-scale
Resolution (Temperature) 0.1
Accuracy (Temperature) ±0.1°C
Display Type LCD
Certifications/Compliance CE
IP Rating IP67
Weight (Metric) 0.6 kg
Warranty 3 Years
Model Cond 3110
Power Supply 4 x 1.5V AA oder 4 x 1.2V NiMH-rechargeable battery
Temperature (Metric) Operating -5°C to +105°C
Type Conductivity Meter Field Kit
Width (Metric) 17.7 cm
Height (Metric) 7.9 cm
Depth (Metric) 5.7 cm
Battery Type NiMH
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.