missing translation for 'onlineSavingsMsg'
Learn More

WTW™ WTW™ ProfiLine™ pH Portable Meters
GREENER_CHOICE

Product Code. 10703774
Change view
Click to view available options
Type:
pH Meter with Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
10703774 pH Meter with Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10703774 Supplier WTW™ Supplier No. 2AA112
  • ONLINE EXCLUSIVE PRICE
    826.00 €
    816.00 € / Pack of 1
    Save 10.00 €
    1% Off
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Obtain optimum results with this easy and robust portable pH/mV meter with an automatic autoread function for routine pH measurements that is safe, secure, reliable, and prevents errors.

  • Clear keypad with only 6 keys
  • Automatic AutoRead function for repeatable measured values make pH measurement safe and prevent errors
  • Anti-skidding keypad that can be operated with gloves
  • Large display that is clear and easy to read
  • Easy and robust portable pH/mV meter for routine measurement
  • Secure and reliable pH measurement due to repeatable results
  • Waterproof according IP 67, 3-point calibration, integrated calibration timer
TRUSTED_SUSTAINABILITY

Specifications

Includes Carry case with pH electrode SenTix™ 41, Buffer solution STP 4 and STP 7, stand, beaker, short instruction manual, CD-ROM and batteries
Accuracy (Temperature) ±0.1°C
Accuracy (mV) ±0.3, ±1
Accuracy (pH) ±0.1, ±0.01, ±0.005
For Use With (Application) Ideal for all seeking an easy, robust and waterproof instrument for portable pH measuring
Range (pH) -2 to 19.999
Resolution (pH) ±0.005
Range (mV) -1200 to 1200, -2000 to 2000
Resolution (mV) ±0.3, ±1mV
Calibration Points 1, 2 or 3 Calibration WTW Technical Buffer, DIN/NIST buffers
Display Type LCD, 7 Segment
Certifications/Compliance CE
IP Rating IP67
Dimensions (L x W x H) 80 W x 180 H mm
Weight (Metric) 0.6 kg
Warranty 3 Years
Model pH 3110 SET 2
Power Supply 4 x 1.5V AA or 4 x 1.2V NiMH rechargeable
Resolution All values ±1 digit
Temperature (Metric) Operating -5°C to +105°C
Battery Type AA, NiMH
Type pH Meter with Kit
Battery Life 2500 hr.
Dimensions (L x W) 80 W mm
Width (Metric) 80 mm
Height (Metric) 180 mm
Depth (Metric) 55 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.