missing translation for 'onlineSavingsMsg'
Learn More

WTW™ ProfiLine™ pH/ION 3310 Pocket Meter

Product Code. 16424540
Change view
Click to view available options
Range (pH):
-2.000 to 19.999
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range (pH) Unit Size
16424540 -2.000 to 19.999 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16424540 Supplier WTW™ Supplier No. 2GA310
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The WTW™ ProfiLine™ pH/ION 3310 is a precise, portable pH/mV/ISE pocket meter with lit background graphic display and manual memory for safe and convenient measurements.

  • The pH 3310 is an elegant combination of portable meter and data logger, with logging intervals between 1 s and 60 minutes
  • The meter includes a built-in data memory for 5000 GLP-compliant entries with an interval-controlled logger
  • Easy 1 to 5-point calibration with calibration timer for all measuring tasks and 22 stored buffer sets
  • Data can conveniently be transmitted to a computer via the USB interface
  • CMC function visualizes the optimum measuring range and thus supports correct measurements
  • Graphic display with clear text menus allows comfortable and safe operation
  • Includes the MultiLab™ Importer to capture data via Excel™
TRUSTED_SUSTAINABILITY

Specifications

Accuracy (mV) ± 0.3 mV/ ± 1 mV
Accuracy (pH) ± 0.005/ ± 0.01/0.1
Accuracy (Temperature) ± 0.1 K
Backlight Option Yes
Calibration Simple calibration with 22 stored buffer sets
Calibration Points 1 to 5-point calibration
Data Management Details Data memory for 5000 GLP-compliant entries
Data Points Data logger with intervals between 1 s and 60 minutes
Measurement Modes pH/mV/Temp
Range (mV) -1250.0 to +1250 mV and -2500 to +2500 mV
Range (pH) -2.000 to 19.999
Temperature (Metric) Range -5.0°C to +105.0°C
Display Type LCD graphic
Model ProfiLine pH/ION 3310
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.