missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Proflavine hemisulfate

Product Code. 16488476
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488476 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16488476 Supplier MedChemExpress Supplier No. HYB0883
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Proflavine hemisulfate, an acridine dye, is a known DNA intercalating agent. Anti-microbial agent.

Proflavine hemisulfate behaves as a pore blocker for Kir3.2. Proflavine hemisulfate is a potential lead compound for Kir3.2-associated neurological diseases.

Specifications

Content And Storage 4°C, sealed storage, away from moisture ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture)
Product Type Dye Reagents
Quantity 100 mg
Shipping Condition Room temperature
CAS 1811-28-5
Molecular Weight (g/mol) 258.29 g/mol
Synonym Proflavin hemisulfate ∣ 3,6-Diaminoacridine hemisulfate
Target HBV
Solubility H2O : ≥ 5 mg/mL (19.36 mM)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.