missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Proflavine hydrochloride

Product Code. 16607052 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 16607052

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 952-23-8 | C26H26Cl2N6O | 509.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 952-23-8
Molecular Formula C26H26Cl2N6O
Molecular Weight (g/mol) 509.44
MDL Number MFCD00066867
InChI Key APOVFGFHTIKDOX-UHFFFAOYSA-N
IUPAC Name dihydrogen bis(acridine-3,6-diamine) hydrate dichloride
SMILES [H+].[H+].O.[Cl-].[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1

Specifications

Alpha Vector PROFLAVINEHYDROCHLORIDE
Melting Point 270°C
Color Brown-Red to Orange
Loss on Drying ≤4 % (105°C, 1 hr)
Infrared Spectrum Conforms
Assay Percent Range 13.6 to 15.0 % (CL) (Titration Argentometric)
Packaging glass bottle
Quantity 5 g
Solubility Information (1 mg/ml aq. soln.) Clear solution
Formula Weight 245.71
Physical Form Powder
Chemical Name or Material Proflavine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Proflavine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Proflavine monohydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.