missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Progesterone

Product Code. 18065572
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18065572 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18065572 Supplier Tocris Bioscience™ Supplier No. 2835/100
  • ONLINE EXCLUSIVE PRICE
    136.00 €
    128.10 € / 100mg
    Save 7.90 €
    5% Off
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Endogenous progesterone receptor agonist

Endogenous progesterone receptor (PR) agonist (EC50 = 0.5 nM).
TRUSTED_SUSTAINABILITY

Specifications

CAS 57-83-0
Quantity 100 mg
Molecular Formula C21H30O2
Formula Weight 314.46
Purity >99%
Molecular Weight (g/mol) 314.469
Chemical Name or Material Progesterone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.