Learn More
DiscoverX ProLinkTM Cloning Vector Bundle
Description
The ProLinkTM Cloning Vector Bundle is a bundle of four vectors for use in expressing a protein of interest fused to variations of the ProLink (PK) tag at the C-terminus. The PK tag is a version of ED used for EFC assays (in combination with EA). The ProLink 2 tag has a higher affinity for EA than ProLink 1, making it ideal for use with weaker protein:protein interactions.The ARMS (Arrestin Recruitment Modifying Sequence) PK tags contain GRK phosphorylation sites to improve ligand-dependent arrestin recruitment to GPCRs and improve assay performance. Provided as four vials, 1 vial each vector, of 10μg vector in 20μL liquid buffer.
Specifications
Specifications
| Product Type | Cloning Vector |
| Quantity | 4 Vectors |
| For Use With (Application) | Cloning Vectors |