missing translation for 'onlineSavingsMsg'
Learn More

Promazine Sulfoxide, TRC

Product Code. 17922585
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17922585 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17922585 Supplier Toronto Research Chemicals Supplier No. TRCP7563105MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 146-21-4
Molecular Formula C17 H20 N2 O S
Quantity 5 mg
Synonym 3-(10H-Phenothiazin-10-yl)-N,N-dimethylpropan-1-amine S-Oxide, Promazine Hydrochloride Imp. A (EP), Promazine Sulphoxide
InChI Formula InChI=1S/C17H20N2OS/c1-18(2)12-7-13-19-14-8-3-5-10-16(14)21(20)17-11-6-4-9-15(17)19/h3-6,8-11H,7,12-13H2,1-2H3
SMILES CN(C)CCCN1c2ccccc2S(=O)c3ccccc13
IUPAC Name N,N-dimethyl-3-(5-oxophenothiazin-10-yl)propan-1-amine
Molecular Weight (g/mol) 300.42
Formula Weight 300.1296
Chemical Name or Material Promazine Sulphoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.