missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Promethazine-d4 HCl (phenothiazine-1,3,7,9-d4), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Promethazine-D4 (phenothiazine-1,3,7,9-D4) Hydrochloride |
| CAS | 1173018-74-0 |
| Molecular Formula | C17H17D4ClN2S |
| Quantity | 0.005 g |
| Synonym | N,N-Dimethyl-1-(1,3,7,9-tetradeuteriophenothiazin-10-yl)propan-2-amine hydrochloride |
| InChI Formula | InChI=1 S/C17H20N2S.ClH/c1-13(18(2)3)12-19-14-8-4-6-10-16(14)20-17-11-7-5-9-15(17)19;/h4-11,13 H,12H2,1-3H3;1 H/i6D,7 D,8 D,9 D; |
| SMILES | [2 H]C1=C2C(SC(C=C([2 H])C=C3[2 H])=C3N2CC(N(C)C)C)=CC([2 H])=C1.Cl |
| IUPAC Name | N,N-dimethyl-1-(10 H-phenothiazin-10-yl-1,3,7,9-d4)propan-2-amine hydrochloride |
| Molecular Weight (g/mol) | 324.9 |
| Formula Weight | 324.14 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction