missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Propafenone-d5 HCl (n-propyl-2,2,3,3,3-d5), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Propafenone 2,2,3,3,3-propyl-d5 Hydrochloride |
| CAS | 1398066-02-8 |
| Molecular Formula | C21 D5 H22 N O3 . Cl H |
| Quantity | 0.005 g |
| Synonym | 1-[2-[2-Hydroxy-3-(propylamino)propoxy]phenyl]-3-phenyl-d5-1-propanone Hydrochloride, 1-[2-[2-Hydroxy-3-(propyl-2,2,3,3,3-d5-amino)propoxy]phenyl]-3-phenyl-1-propanone hydrochloride, 1-[2-[2-Hydroxy-3-(pentadeuteriopropylamino)propoxy]phenyl]-3-phenylpropan-1-one hydrochloride, 2'-[2-Hydroxy-3-(propylamino)propoxy]-3-phenyl-d5-propiophenone Hydrochloride, Arythmol-d5, Pronon-d5, Rythmol-d5, Rytmonorm-d5, Propafenone-D5 (propyl-2,2,3,3,3-D5) hydrochloride, Propafenone-d5 hydrochloride |
| InChI Formula | InChI=1 S/C21H27NO3.ClH/c1-2-14-22-15-18(23)16-25-21-11-7-6-10-19(21)20(24)13-12-17-8-4-3-5-9-17;/h3-11,18,22-23 H,2,12-16H2,1H3;1 H/i1D3,2D2; |
| SMILES | Cl.[2 H]C([2 H])([2 H])C([2 H])([2 H])CNCC(O)COc1ccccc1C(=O)CCc2ccccc2 |
| IUPAC Name | 1-[2-[2-hydroxy-3-(2,2,3,3,3-pentadeuteriopropylamino)propoxy]phenyl]-3-phenylpropan-1-one;hydrochloride |
| Molecular Weight (g/mol) | 382.94 |
| Formula Weight | 382.2072 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction