missing translation for 'onlineSavingsMsg'
Learn More

Propanol-2 (iso-propanol), HPLC Grade For Laboratory Use and HPLC, Chem Lab

Product Code. 15969286
Click to view available options
:
Propanol-2 (iso-propanol), HPLC Grade For Laboratory Use and HPLC, 2.5 L
This item is not returnable. View return policy

Product Code. 15969286

Brand: Chem-Lab CL00.0920.2500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Melting Point -89°C
CAS 67-63-0
Color Colorless
Acidity < 0.0005 meq/g
Density 0.78 g/mL
Boiling Point 82°C
Quantity 2.5 L
Flash Point 12 °C
Packaging Glass Bottle/H
Physical Form Liquid
Assay Percent Range > 99.8%
Molecular Formula C3H8O
Water < 1000 ppm
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILES CC(C)O
IUPAC Name propan-2-ol
Molecular Weight (g/mol) 60.10
Grade HPLC
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : iso-Propanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.