missing translation for 'onlineSavingsMsg'
Learn More

Propargyl acetate, 97%

Product Code. 10741714 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10741714 25 g Glass bottle 25g
10646114 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10741714 Supplier Thermo Scientific Acros Supplier No. 380820250
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 627-09-8 | C5H6O2 | 98.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-09-8
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.1
MDL Number MFCD00041601
InChI Key RIZZXCJMFIGMON-UHFFFAOYSA-N
Synonym propargyl acetate, 2-propyn-1-ol, acetate, 3-acetoxypropyne, 2-propynyl acetate, propargyl alcohol acetate, prop-2-yn-1-yl acetate, acmc-1awj3, 2-propyn-1-ol, 1-acetate, 4-02-00-00197 beilstein handbook reference
PubChem CID 69388
IUPAC Name prop-2-ynyl acetate
SMILES CC(=O)OCC#C

Specifications

Color Colorless
Density 0.9890g/mL
Boiling Point 27.0°C to 28.0°C (8.0 mmHg)
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH≡CCH2OCOCH3
Refractive Index 1.4160 to 1.4180
Quantity 25 g
Specific Gravity 0.989
Formula Weight 98.1
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Propargyl acetate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Propargyl acetate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.