missing translation for 'onlineSavingsMsg'
Learn More

Propargyl acrylate, 96%, stab. with ca 200ppm BHT

Product Code. 11377396
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377396 10 g 10g
11367396 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377396 Supplier Thermo Scientific Alfa Aesar Supplier No. L09383.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10477-47-1 | C6H6O2 | 110.11 g/mol

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10477-47-1
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.11
MDL Number MFCD00078451
InChI Key WPBNLDNIZUGLJL-UHFFFAOYSA-N
Synonym propargyl acrylate, 2-propenoic acid, 2-propynyl ester, acrylic acid propargyl ester, 2-propenoic acid, 2-propyn-1-yl ester, 2-propynyl acrylate, prop-2-yn-1-yl acrylate, acmc-1c257, prop-2-yn-1-yl prop-2-enoate, propargyl acrylate,
PubChem CID 82655
SMILES C=CC(=O)OCC#C

Specifications

Density 0.995
Boiling Point 138°C
Flash Point 43°C (109°F)
Odor Pungent
Linear Formula H2C=CHCO2CH2C≡CH
Refractive Index 1.446
Quantity 10 g
UN Number UN3272
Beilstein 1902961
Solubility Information Not miscible or difficult to mix with water.
IUPAC Name prop-2-ynyl prop-2-enoate
Formula Weight 110.11
Percent Purity 96%
Chemical Name or Material Propargyl acrylate, stabilized with ≈200ppm BHT
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.