missing translation for 'onlineSavingsMsg'
Learn More

Propargyl Alcohol, TRC

Product Code. 30415295
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30415295 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30415295 Supplier Toronto Research Chemicals Supplier No. TRCP76255050ML

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Propargyl alcohol
CAS 107-19-7
Molecular Formula C3 H4 O
Quantity 50 mL
Synonym 1-Hydroxy-2-propyne, 1-Propyn-3-ol, 1-Propyn-3-yl alcohol, 2-Propynol, 2-Propynyl alcohol, 3-Hydroxy-1-propyne, 3-Hydroxypropyne, 3-Propynol, Ethynylcarbinol, Hydroxymethylacetylene, NSC 8804, Propargyl alcohol, Propynyl alcohol
InChI Formula InChI=1S/C3H4O/c1-2-3-4/h1,4H,3H2
SMILES OCC#C
IUPAC Name prop-2-yn-1-ol
Molecular Weight (g/mol) 56.06
Formula Weight 56.0262
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.